Anti-PRDM2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-6062R-A750
: Bioss
- Antibody type:Primary
- Antigen name:PRDM2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:1301-1400/1718
- Form:Liquid
- Gene ID:7799
- Antigen synonyms:HUMHOXY 1|RIZ 2|Retinoblastoma protein interacting zinc- inger protein|PR domain containing protein 2|RIZ2|Zinc finger protein RIZ|retinoblastoma protein-interacting zinc finger protein|PR domain containing 2 with ZNF domain|RIZ 1|PR domain-containing protein 2|MTB-ZF|MTB ZF|Lysine N-methyltransferase 8|Lysine N methyltransferase 8|RIZ-LSB|PRDM2|RIZ1|RIZ|PR domain zinc finger protein 2|GATA-3-binding protein G3B|PRDM2_HUMAN|PRDM 2|GATAGATA 3 binding protein G3B|Retinoblastoma protein-interacting zinc-finger protein|Retinoblastoma protein binding zinc finger protein|HUMHOXY1|MTE-binding protein|Zinc finger DNA binding protein|MTE binding protein|PR-domain zinc finger protein 2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:189 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PRDM2/RIZ1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
S-adenosyl-L-methionine-dependent histone methyltransferase that specifically methylates 'Lys-9' of histone H3. May function as a DNA-binding transcription factor. Binds to the macrophage-specific TPA-responsive element (MTE) of the HMOX1 (heme oxygenase 1) gene and may act as a transcriptional activator of this gene.
Type: Primary
Antigen: PRDM2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1301-1400/1718
Host: Rabbit
Isotype: IgG
Reactivity: