Anti-MCL1 Rabbit Polyclonal Antibody
ROCK200-401-CS0
- Antibody type:Primary
- Antigen name:Induced myeloid leukemia cell differentiation protein Mcl-1
- Antigen symbol:MCL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Format:IgG
- Form:Liquid (sterile filtered)
- Gene ID:4170
- Antigen synonyms:BCL2L3|MCL1L|Bcl-2-like protein 3|Mcl-1|mcl1/EAT|Induced myeloid leukemia cell differentiation protein Mcl-1|bcl2-L-3|MCL1S|Bcl2-L-3|TM|MCL1-ES|EAT
- Storage buffer:0.01 M Sodium Phosphate, 0.25 M Sodium Chloride, pH 7.2
- Storage temperature:-20°C
- Concentration:1 mg/mL by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:Anti-Mcl-1 antibody was prepared from whole rabbit serum produced by repeated immunizations with a peptide corresponding to 16 amino acids near the C-terminus of human Mcl-1.
- Pk:100 µG