Order Entry
Italy
ContactUsLinkComponent
Anti-CSTF3/CSTF77 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-CSTF3/CSTF77 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-12935R-A750
Supplier:  Bioss
Anti-CSTF3/CSTF77 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-12935R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CSTF3/CSTF77
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    151-250/717
  • Form:
    Liquid
  • Gene ID:
    1479
  • Antigen synonyms:
    MGC75122|CSTF77|CSTF 77 kDa subunit|CSTF 3|Cleavage stimulation factor subunit 3|Cleavage stimulation factor 77 kDa subunit|MGC117398|CSTF3_HUMAN|CF 1 77 kDa subunit|Cleavage stimulation factor subunit 3 isoform 1|MGC43001|CSTF 77
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    83 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human CSTF3/CSTF77
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Polyadenylation of mRNA precursors is a two-step reaction that requires multiple protein factors. The first step, endonucleolytic cleavage of polyadenylation substrates, requires CstF (cleavage stimulation factor), a heterotrimer that is composed of three distinct subunits. Heterotrimeric CstF recognizes GU- and U-rich sequences located downstream of the polyadenylation site on RNA. CstF-77 (cleavage stimulation factor, 77 kDa subunit), also known as CstF3, is one of the three subunits comprising CstF. It can exist as a homodimer and functions as the bridge, directly interacting with the other two CstF subunits, namely CstF-64 and CstF-50. CstF-77 is highly conserved among eukaryotes. It contains an Alpha-helical structure with 11 HAT (Half-a-TPR-containing) repeats and is essential for CstF assembly. In addition, CstF-77 is capable of interacting with CPSF1 and FCP1, other factors involved in polyadenylation.

Type: Primary
Antigen: CSTF3/CSTF77
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-250/717
Host: Rabbit
Isotype: IgG
Reactivity: Mouse