Anti-COX6C Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-12925R-A750
: Bioss
- Antibody type:Primary
- Antigen name:COX6c/Cytochrome C Oxidase subunit VIc
- Antigen symbol:COX6C
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:21-75/75
- Form:Liquid
- Gene ID:1345
- Antigen synonyms:Cytochrome c oxidase polypeptide VIc|COX6C|Cytochrome c oxidase subunit 6C
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:9 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human COX6c/Cytochrome C Oxidase subunit VIc
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
Type: Primary
Antigen: COX6c/Cytochrome C Oxidase subunit VIc
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-75/75
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse