Specifications
- Antibody type:Primary
- Antigen name:ospA
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Bacteria
- Epitope:1-100/273
- Form:Liquid
- Antigen synonyms:Borreliella afzelii|Borrelia mayonii|Lyme disease|Borrelia|Borreliella yangtze|Borreliella bavariensis|Borreliella chilensis|Borreliella valaisiana|Borreliella finlandensis|Borreliella japonica|Borreliaceae|ospA|Lyme disease spirochete|Borrelia lanei|Borreliella burgdorferi|Borreliella californiensis|Borreliella lusitaniae|Borreliella spielmanii|Borreliella bissettii|Borreliella garinii
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:83 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from Borrelia burgdorferi Outer surface protein A
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Borrelia burgdorferi is a spirochete and the cause of Lyme disease, a tick-transmitted illness of humans and animals. B. burgdorferi may persist in humans and animals for months or years following initial infection, despite a robust humoral immune response. B. burgdorferi resembles other spirochetes in that it is a highly specialised, motile, two-membrane, spiral shaped bacteria which lives primarily as an extracellular pathogen. B. burgdorferi has an unusual genome compared with other eubacteria which includes a linear chromosome approximately one megabase in size and numerous linear and circular plasmids.
Type: Primary
Antigen: ospA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/273
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria