Specifications
- Antibody type:Primary
- Antigen name:TCTN3/TECT3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:201-300/607
- Form:Liquid
- Gene ID:26123
- Antigen synonyms:TECT3_HUMAN|DKFZP564D116|Tectonic family member 3|Tectonic 3 precursor|C10orf61|Tectonic-3|Chromosome 10 open reading frame 61|TCTN3|Tectonic 3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:64 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TCTN3/TECT3
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Tect3 is a 607 amino acid single-pass type I membrane protein that belongs to the tectonic family and exists as four alternatively spliced isoforms. Tect3 interacts with MKS1 and may be involved in apoptosis regulation. The gene that encodes Tect3 contains approximately 31560 bases and maps to human chromosome 10q24.1. Spanning nearly 135 million base pairs and encoding nearly 1200 genes, chromosome 10 makes up approximately 4.5% of the human genome. Several protein-coding genes, including those that encode chemokines, cadherins, excision repair proteins, early growth response factors (Egrs) and fibroblast growth receptors (FGFRs), are located on chromosome 10. Defects in some of the genes that map to chromosome 10 are associated with Charcot-Marie Tooth disease, Jackson-Weiss syndrome, Usher syndrome, nonsyndromatic deafness, Wolman’s syndrome, Cowden syndrome, Cockayne syndrome, multiple endocrine neoplasia type 2 and porphyria. Tetrahydrobiopterin deficiency and a number of syndromes involving defective skull and facial bone fusion are also linked to chromosome 10.
Type: Primary
Antigen: TCTN3/TECT3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/607
Host: Rabbit
Isotype: IgG
Reactivity: