Order Entry
Italy
ContactUsLinkComponent
Anti-ENSA/ARPP-19 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-ENSA/ARPP-19 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-11793R-A750
Supplier:  Bioss
Anti-ENSA/ARPP-19 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-11793R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ENSA/ARPP-19
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    51-121/121
  • Form:
    Liquid
  • Antigen synonyms:
    ARPP 19e|Alpha endosulfine|ENSA_HUMAN|Endosulfine alpha
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    13 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human ENSA/ARPP-19
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Sulfonylureas are a class of drugs commonly used in the management of noninsulin-dependent diabetes mellitus. ENSA/ARPP-19, also known as ARPP-19e, is a 121 amino acid protein that localizes to the cytoplasm and belongs to the endosulfine family. ENSA/ARPP-19 is an endogenous regulator of the cell K(ATP) channels and reduces K(ATP) channel currents by inhibiting sulfonylurea from binding to the receptor thereby stimulating insulin secretion by pancreatic cells. It is suggested that ENSA/ARPP-19 expression is decreased in brains of both Alzheimer's disease (AD) and Down's syndrome patients. Widely expressed with high levels in skeletal muscle and brain and lower levels in pancreas, ENSA/ARPP-19 is phosphorylated by PKA and exists as eight alternatively spliced isoforms. ENSA/ARPP-19 is encoded by a gene located on human chromosome 1q21.3.

Type: Primary
Antigen: ENSA/ARPP-19
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-121/121
Host: Rabbit
Isotype: IgG
Reactivity: