Specifications
- Antibody type:Primary
- Antigen name:Neuron navigator 1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:1426-1475/1877
- Form:Liquid
- Antigen synonyms:Steerin 1|Protein Steerin 1|POMFIL3|Protein Steerin1|Nav1|Steerin1|mNav1|unc-53 homolog 1|FLJ14203|DKFZp781D0314|Pore membrane and/or filament interacting like protein 3|FLJ12560|Nav 1|NAV1_HUMAN|POMFIL 3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:202 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Neuron navigator 1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Neuron navigator 1 is a 1877 amino acid cytoplasmic protein that is involved in neuronal migration. Neuron navigator 1 is widely expressed at low levels, though highest expression is found in both adult and fetal nervous tissue. Through interaction with tubulin, Neuron navigator 1 associates with a subset of mirotubule plus ends present in the growth cone. Overexpression of Neuron navigator 1 leads to microtubule bundling, whereas a reduction of its levels causes loss of directionality in the migration of pontine cell leading processes. There are seven isoforms of Neuron navigator 1 that are produced as a result of alternative splicing events.
Type: Primary
Antigen: Neuron navigator 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1426-1475/1877
Host: Rabbit
Isotype: IgG
Reactivity: