Anti-Synaptotagmin 10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-11400R-A750
: Bioss
- Antibody type:Primary
- Antigen name:Synaptotagmin 10
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:351-435/523
- Form:Liquid
- Antigen synonyms:SYT10_HUMAN|Synaptotagmin-10|Synaptotagmin 10|SynaptotagminX|Synaptotagmin-X|SYT-10|SYT 10|SYT10|Synaptotagmin10|SytX
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:59 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Synaptotagmin X
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
May be involved in Ca²⁺ dependent exocytosis of secretory vesicles through Ca²⁺ and phospholipid binding to the C2 domain or may serve as Ca²⁺ sensors in the process of vesicular trafficking and exocytosis.
Type: Primary
Antigen: Synaptotagmin 10
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 351-435/523
Host: Rabbit
Isotype: IgG
Reactivity: