Order Entry
Italy
ContactUsLinkComponent
Anti-C20orf160 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-C20orf160 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-9695R-A680
 :  Bioss
Anti-C20orf160 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-9695R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    C20orf160
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    41-140/571
  • Form:
    Liquid
  • Gene ID:
    140706
  • Antigen synonyms:
    FLJ43600|chromosome 20 open reading frame 160|CCM2L_HUMAN|hypothetical protein LOC140706|dJ310O13.5
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human C20orf160
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Representing about 2% of human DNA, chromosome 20 consists of approximately 63 million bases and 600 genes. Chromosome 20 contains a region with numerous genes expressed in the epididymis, which are thought important for seminal production, and some viewed as potential targets for male contraception. The PRNP gene encoding the prion protein associated with spongiform encephalopathies, like Creutzfeldt-Jakob disease, is found on chromosome 20. Amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome are also associated with chromosome 20. The C20orf160 gene product has been provisionally designated C20orf160 pending further characterisation.

Type: Primary
Antigen: C20orf160
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 41-140/571
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat