Anti-factor 13 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-1587R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:factor 13
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:331-732/732
- Form:Liquid
- Gene ID:2162
- Antigen synonyms:Transglutaminase A chain|Protein-glutamine gamma-glutamyltransferase A chain|Coagulation factor XIIIa|F13A|Coagulation factor XIII A chain|F13A1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:76 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human factor XIII
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Factor XIII is activated by thrombin and calcium ion to a transglutaminase that catalyses the formation of gamma-glutamyl-epsilon-lysine cross-links between fibrin chains, thus stabilizing the fibrin clot. Also cross-link alpha-2-plasmin inhibitor, or fibronectin, to the alpha chains of fibrin.
Type: Primary
Antigen: factor 13
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 331-732/732
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse