Anti-PUMA Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-1573R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:PUMA
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Rat
- Western blot:Yes
- Epitope:131-180/193
- Form:Liquid
- Gene ID:27113
- Antigen synonyms:BBC3|p53 up-regulated modulator of apoptosis|JFY-1|PUMA|JFY1|Bcl-2-binding component 3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:21 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PUMA
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Essential mediator of p53/TP53-dependent and p53/TP53-independent apoptosis. Isoform 3 fails to show any growth-inhibitory or apoptotic activity.
Type: Primary
Antigen: PUMA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 131-180/193
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Pig