Specifications
- Antibody type:Primary
- Antigen name:Ghrelin 28
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:24-51/117
- Form:Liquid
- Gene ID:58991
- Antigen synonyms:motilin-related peptide|grehlin|growth hormone secretagogue|protein M46|growth hormone-releasing peptide|Appetite-regulating hormone
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:13 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide of mouse Ghrelin
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Ghrelin is an endogenous ligand for the growth hormone secretagogue receptor and is involved in regulating growth hormone release. Ghrelin is derived from a preprohormone called preproghrelin, which also generates a second peptide called obestatin. Obestatin is an endogenous ligand for the orphan G protein-coupled receptor GPR39 and is involved in satiety and decreased food intake. Also known as Appetite regulating hormone; GHRL; Growth hormone releasing peptide; Growth hormone secretagogue; M46 protein; Motilin related peptide; MTLRP; Obestatin; Obestatin preprohormone; PRO1066; UNQ524. Sequence notes: Gly-Ser-Ser-Phe-Leu-Ser-Pro- Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu- Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro- Arg (mo, rat).
Type: Primary
Antigen: Ghrelin 28
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 24-51/117
Host: Rabbit
Isotype: IgG
Reactivity: Rat