Order Entry
Italy
ContactUsLinkComponent
Anti-SRC Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-SRC Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-1135R-A680
 :  Bioss
Anti-SRC Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-1135R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    SRC
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    2-100/541
  • Form:
    Liquid
  • Gene ID:
    6714
  • Antigen synonyms:
    c SRC|ASV|Oncogene SRC|protooncogene SRC Rous sarcoma|Neuronal SRC|Src|Neuronal CSRC tyrosine specic protein kinase|pp60c src|p60 Src|Tyrosine protein kinase SRC 1|SRC Oncogene|p60-Src|SRC_HUMAN.|Tyrosine kinase pp60c src|pp60csrc|V src sarcoma Schmidt Ruppin A 2 viral oncogene homolog avian|Proto-oncogene c-Src|SRC_MOUSE|p60Src|CDNA FLJ14219 fis clone NT2RP3003800 highly similar to Rattus norvegicus tyrosine protein kinase pp60 c src mRNA|Protooncogene SRC|OTTHUMP00000174476|Avian sarcoma virus|vsrc avian sarcoma Schmidt Ruppin A2 viral oncogene homolog|v src sarcoma Schmidt Ruppin A 2 viral oncogene homolog avian|pp60c-src|SRC1|Proto oncogene tyrosine protein kinase Src|OTTHUMP00000174477|Proto-oncogene tyrosine-protein kinase Src|Tyrosine protein kinase SRC1|v src avian sarcoma Schmidt Ruppin A2 viral oncogene homolog|EC 2.7.10.2|OTTHUMP00000030931|cSrc
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    61 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human SRC
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

c-Src tyrosine kinase plays a critical role in signal transduction downstream of growth factor receptors, integrins and G protein-coupled receptors. We used stable isotope labelling with amino acids in cell culture (SILAC) approach to identify additional substrates of c-Src tyrosine kinase in human embryonic kidney 293T cells. We have identified 10 known substrates and interactors of c-Src and Src family kinases along with 26 novel substrates. We have experimentally validated 4 of the novel proteins (NICE-4, RNA binding motif 10, FUSE-binding protein 1 and TRK-fused gene) as direct substrates of c-Src using in vitro kinase assays and cotransfection experiments. Significantly, using a c-Src specific inhibitor, we were also able to implicate 3 novel substrates (RNA binding motif 10, EWS1 and Bcl-2 associated transcription factor) in PDGF signaling. Finally, to identify the exact tyrosine residues that are phosphorylated by c-Src on the novel c-Src substrates, we designed custom peptide microarrays containing all possible tyrosine-containing peptides (312 unique peptides) and their mutant counterparts containing a Tyr --> Phe substitution from 14 of the identified substrates. Using this platform, we identified 34 peptides that are phosphorylated by c-Src. We have demonstrated that SILAC-based quantitative proteomics approach is suitable for identification of substrates of nonreceptor tyrosine kinases and can be coupled with peptide microarrays for high-throughput identification of substrate phosphopeptides.

Type: Primary
Antigen: SRC
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 2-100/541
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat