Order Entry
Italy
ContactUsLinkComponent
Anti-ACTH 1-39 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-ACTH 1-39 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0443R-A750
Supplier:  Bioss
Anti-ACTH 1-39 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0443R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ACTH 1-39
  • Antigen symbol:
    ACTH 1-39
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Epitope:
    1-39/39
  • Form:
    Liquid
  • Gene ID:
    5443
  • Antigen synonyms:
    MSH|NPP|POC|CLIP|LPH|Corticotropin-lipotropin|POMC|ACTH|Pro-opiomelanocortin
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    4.5 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human ACTH
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

ACTH stimulates the adrenal glands to release cortisol. MSH (melanocyte-stimulating hormone) increases the pigmentation of skin by increasing melanin production in melanocytes. Beta-endorphin and Met-enkephalin are endogenous opiates.

Type: Primary
Antigen: ACTH 1-39
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-39/39
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat