Anti-PTK9 Tyr321 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-5613R-A680
: Bioss
- Antibody type:Primary
- Antigen name:PTK9 Tyr321
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:5756
- Antigen synonyms:A6|Twinfilin 1|PTK9 Tyr321|PTK9 Y321|TWF 1|Twinfilin|PTK9 protein tyrosine kinase 9|TWF1|p-PTK9 Tyr321|TWF1_HUMAN|Protein tyrosine kinase 9|PTK9|Twinfilin, Drosophila, homolog of, 1|p-TWF1Y321|Twinfilin actin binding protein, homolog 1|Twinfilin-1|A6 protein tyrosine kinase|Protein A6|Twinfilin, actin-binding protein, homolog 1 Drosophila
- Modification:Phosphorylation (Tyr321)
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:40 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human TWF1 around the phosphorylation site of Tyr321
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
PTK9 is an actin-binding protein involved in motile and morphological processes. It inhibits actin polymerization, likely by sequestering G-actin. By capping the barbed ends of filaments, it also regulates motility. PTK9 seems to play an important role in clathrin-mediated endocytosis and distribution of endocytic organelles.
Type: Primary
Antigen: PTK9 Tyr321
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: