Anti-PYGL Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-5011R-A680
: Bioss
- Antibody type:Primary
- Antigen name:PYGL
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:451-550/847
- Form:Liquid
- Gene ID:5836
- Antigen synonyms:GSD6 antibody Hers disease, glycogen storage disease type VI|Phosphorylase glycogen liver|Glycogen phosphorylase liver form|PYGL_HUMAN|Glycogen phosphorylase liver
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:97 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PYGL
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Phosphorylase is an important allosteric enzyme in carbohydrate metabolism. Enzymes from different sources differ in their regulatory mechanisms and in their natural substrates. However, all known phosphorylases share catalytic and structural properties.
Type: Primary
Antigen: PYGL
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 451-550/847
Host: Rabbit
Isotype: IgG
Reactivity: