Anti-CD24 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-4891R-A750
: Bioss
- Antibody type:Primary
- Antigen name:CD24
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:27-53/76
- Form:Liquid
- Gene ID:12484
- Antigen synonyms:R13-Ag|Cd24a|Ly-52|HSA|Lymphocyte antigen 52|nectadrin|M1/69-J11D heat stable antigen|Cd24|X62 heat stable antigen|Signal transducer CD24
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:8 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse CD24
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
May have a specific role to play in early thymocyte development.
Type: Primary
Antigen: CD24
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 27-53/76
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat