Anti-SV2A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2407R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:SV2A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Epitope:451-550/743
- Form:Liquid
- Gene ID:9900
- Antigen synonyms:PSEC0174|Synaptic vesicle glycoprotein 2A|SV2|KIAA0736|SV2A
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:83 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SV2A
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Plays a role in the control of regulated secretion in neural and endocrine cells, enhancing selectively low-frequency neurotransmission. Positively regulates vesicle fusion by maintaining the readily releasable pool of secretory vesicles (By similarity).
Type: Primary
Antigen: SV2A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 451-550/743
Host: Rabbit
Isotype: IgG
Reactivity: Human