Order Entry
Italy
ContactUsLinkComponent
Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-2197R-A680
 :  Bioss
Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-2197R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    duck hepatitis A virus 1 Genome polyprotein
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Western blot:
    Yes
  • Epitope:
    851-950/2249
  • Form:
    Liquid
  • Antigen synonyms:
    DHAV|polyprotein DHAV|polyprotein [duck hepatitis A virus]
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    252 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    MAP peptide derived from polyprotein [duck hepatitis A virus] FC64/CHN/2009
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Duck hepatitis A virus genome polyprotein.

Type: Primary
Antigen: duck hepatitis A virus 1 Genome polyprotein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 851-950/2249
Host: Rabbit
Isotype: IgG
Reactivity: Virus; Others