Anti-Melatonin Receptor 1A Rabbit Polyclonal Antibody
Catalog # BOSSBS-20768R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Melatonin Receptor 1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:121-200/350
- Form:Liquid
- Gene ID:4543
- Antigen synonyms:MTNR 1A|MEL-1A-R|MT 1|MelR|Mel1a receptor|MTR-1A|MTR1A_HUMAN|Mel1a melatonin receptor|MT1|Mel 1A R|MR|Melatonin receptor type 1A|Melatonin receptor 1A|MTNR1A
- Modification:Unmodified
- Storage buffer:0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:39 kDa
- Storage temperature:Shipped at 4 °C. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Melatonin Receptor 1A
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
High affinity receptor for melatonin. Likely to mediates the reproductive and circadian actions of melatonin. The activity of this receptor is mediated by pertussis toxin sensitive G proteins that inhibit adenylate cyclase activity.
Type: Primary
Antigen: Melatonin Receptor 1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Public Immunogen Range: 121-200/350
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat