Specifiche
- Antibody type:Primary
- Antigen name:Beta Amyloid 1 - 42
- Clonality:Monoclonal
- Clone:1F4
- Conjugation:Alexa Fluor® 750
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:351
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human beta-Amyloid (1-42)
- Purification:Purified by Protein G
- Pack type:Vial
- Pk:100 µl
Specifiche
Informazioni su questo articolo
The cerebral and vascular plaques associated with Alzheimer's disease are mainly composed of Amyloid beta peptides. beta Amyloid is derived from cleavage of the Amyloid precursor protein and varies in length from 39 to 43 amino acids. beta Amyloid [1-40], beta Amyloid [1-42], and beta Amyloid [1-43] peptides result from cleavage of Amyloid precursor protein after residues 40, 42, and 43, respectively. The cleavage takes place by gamma-secretase during the last Amyloid precursor protein processing step. beta Amyloid [1-40], beta Amyloid [1-42], and beta Amyloid [1-43] peptides are major constituents of the plaques and tangles that occur in Alzheimer's disease. beta Amyloid and peptides have been developed as tools for elucidating the biology of Alzheimer's disease.
Type: Primary
Antigen: beta Amyloid 1-42
Clonality: Monoclonal
Clone: 1F4
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Mouse
Isotype: IgG
Reactivity: Human; Rat