Specifications
- Antibody type:Primary
- Antigen name:IGFBPL1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:201-278/278
- Form:Liquid
- Antigen synonyms:bA113O24,1|insulin-like growth factor-binding-related protein 4|insulin-like growth factor binding protein related protein 4|IGFBPRP4|IGFBP-related protein 10|IBPL1_HUMAN|insulin-like growth factor-binding protein-like 1|IGFBP-RP4
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:26 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human IGFBPL1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
IGFBPL1 is a secreted IGF (Insulin-like growth factor) binding protein that is known to contain an Ig-like C2-type (immunoglobulin-like) domain, an IGFBP N-terminal domain and a Kasal-like domain. IGF-binding proteins characteristically act to extend the half-life of IGFs and may influence the growth promoting effects of the IGFs. The interaction of IGFBPs with IGFs can affect cell surface receptors, specifically, IGFBPs may enhance or decrease a cells insulin sensitivity. IGFBPL1 has been found to be down-regulated in multiple tumors and thus may be a likely tumor suppressor candidate. Highly expressed in both brain and testis, IGFBPL1 is found at lower levels in the prostate, bladder and lung.
Type: Primary
Antigen: IGFBPL1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-278/278
Host: Rabbit
Isotype: IgG
Reactivity: