Anti-HFM1 Rabbit Polyclonal Antibody (FITC)
Catalog # BOSSBS-15471R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:HFM1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:151-250/1435
- Form:Liquid
- Antigen synonyms:HFM1|SEC63 domain-containing protein 1|HFM1_HUMAN|Probable ATP-dependent DNA helicase HFM1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:163 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HFM1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
HFM1.
Type: Primary
Antigen: HFM1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 151-250/1435
Host: Rabbit
Isotype: IgG
Reactivity: