Anti-HERC6 Rabbit Polyclonal Antibody (Cy7)
Catalog # BOSSBS-15463R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:HERC6
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:751-850/1022
- Form:Liquid
- Antigen synonyms:HECT domain and RCC1 like domain containing protein 6|Probable E3 ubiquitin protein ligase HERC6|HERC6_HUMAN|FLJ20637|Probable E3 ubiquitin-protein ligase HERC6|Potential ubiquitin ligase|HERC6|Hect domain and RLD 6|HECT domain and RCC1-like domain-containing protein 6|EC 6,3,2,|HERC 6
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:115 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HERC6
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
HERC6.
Type: Primary
Antigen: HERC6
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 751-850/1022
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat