Anti-HCE Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Catalog # BOSSBS-15425R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:HCE
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:451-550/597
- Form:Liquid
- Antigen synonyms:HCAP1|mRNA guanylyltransferase|HCE1|mRNA guanyltransferase|GTase|mRNA 5''-triphosphatase|mRNA capping enzyme|RNA guanylyltransferase and 5' phosphatase|HcE|TPase|polynucleotide 5' trisphosphatase|CAP1A|MCE1_HUMAN|MCE1|RNGTT|GTP--RNA guanylyltransferase|hCAP
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:69 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HCE
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
HCE.
Type: Primary
Antigen: HCE
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 451-550/597
Host: Rabbit
Isotype: IgG
Reactivity: