Anti-GPCR TAS1R1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)
BOSSBS-15373R-A647
: Bioss
- Antibody type:Primary
- Antigen name:GPCR TAS1R1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:431-530/841
- Form:Liquid
- Antigen synonyms:Taste receptor type 1 member 1|TAS1 R1|Krueppel related zinc finger protein 3 (HKR3 protein)|G protein coupled receptor 70|TAS1R1|TR 1|seven transmembrane helix receptor|TR1|taste receptor, type 1, member 1|gm 148|gm148|TS1R1|TS1R1_HUMAN|Sweet taste receptor T1r|GPR 70|T1R1|G protein-coupled receptor 70|GPR70|G-protein coupled receptor 70
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:91 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GPCR TAS1R1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
GPCR TAS1R1.
Type: Primary
Antigen: GPCR TAS1R1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 431-530/841
Host: Rabbit
Isotype: IgG
Reactivity: