Specifications
- Antibody type:Primary
- Antigen name:C9orf23
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:1-100/163
- Form:Liquid
- Gene ID:138716
- Antigen synonyms:RPP25L|Ribonuclease P protein subunit p25 like protein|RP25L_HUMAN|Alba like protein C9orf23|Ribonuclease P/MRP 25 kDa subunit like|C9orf23|MGC29635|bA296L22,5|Rpp25 like protein|RNase P protein subunit like p25
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:18 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C9ORF23
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
C9orf23 (chromosome 9 open reading frame 23) is a 163 amino acid protein that belongs to the histone-like Alba family and is encoded by a gene that maps to human chromosome 9p13.3. Chromosome 9 consists of about 145 million bases, represents 4% of the human genome and encodes nearly 900 genes. Thought to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterised by harmful vascular defects, is associated with the chromosome 9 gene encoding endoglin protein, ENG. Familial dysautonomia is also associated with chromosome 9 though through the gene IKBKAP. Notably, chromosome 9 encompasses the largest interferon family gene cluster. Chromosome 9 is partnered with chromosome 22 in the translocation leading to the aberrant production of BCR-ABL fusion protein often found in leukemias.
Type: Primary
Antigen: C9orf23
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 1-100/163
Host: Rabbit
Isotype: IgG
Reactivity: