Anti-GTP binding protein REM1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-7502R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:GTP binding protein REM1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:221-298/298
- Form:Liquid
- Gene ID:28954
- Antigen synonyms:E030011C07Rik|REM 1|RAS RAD and GEM like GTP binding|GD:REM|MGC48669|Rad and Gem like GTP binding protein 1|OTTMUSP00000016855|REM1|GTP binding protein REM1|Rad and Gem related GTP binding protein|REM1_HUMAN|GES|GTPase GES|RAS like GTP binding protein REM|GTP binding protein REM|MGC114556|GTPase regulating endothelial cell sprouting|GTP binding protein REM 1|RP23-35I8.9|RAS like GTP binding
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:33 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GTP binding protein REM1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Promotes endothelial cell sprouting and actin cytoskeletal reorganization. May be involved in angiogenesis. May function in Ca2+ signaling.
Type: Primary
Antigen: GTP binding protein REM1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 221-298/298
Host: Rabbit
Isotype: IgG
Reactivity: Mouse