Anti-RIPK3/RIP3 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-77299
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Receptor-interacting serine/threonine-protein kinase 3
- Antigen symbol:RIPK3/RIP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:11035
- Antigen synonyms:RIP-like protein kinase 3|RIP3receptor interacting protein 3|Receptor-interacting protein 3|RIP-3|EC 2.7.11.1|receptor-interacting serine/threonine-protein kinase 3|receptor-interacting serine-threonine kinase 3
- Storage buffer:PBS
- Storage temperature:Store at 4 °C short term. Aliquot and store at ‒20 °C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Antibody was raised against a 14 amino acid peptide near the carboxy terminus of murine RIP3. The immunogen is located within the last 50 amino acids of RIP3.
- Purification:Immunogen affinity purified
- Pk:100 µG
Specifications
About this item
The RIPK3 / RIP3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RIPK3 / RIP3. This antibody reacts with human, mouse, rat. The RIPK3 / RIP3 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 5 µg/ml
Type: Primary
Antigen: RIPK3/RIP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat