Order Entry
Italy
ContactUsLinkComponent
Anti-Adenosine A2a R Mouse Monoclonal Antibody [clone: 7F6-G5-A2]
Anti-Adenosine A2a R Mouse Monoclonal Antibody [clone: 7F6-G5-A2]
Catalog # NOVUNBP1-39474
Supplier:  Novus Biologicals
Anti-Adenosine A2a R Mouse Monoclonal Antibody [clone: 7F6-G5-A2]
Catalog # NOVUNBP1-39474
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Adenosine A2a R
  • Antigen symbol:
    ADORA2
  • Clonality:
    Monoclonal
  • Clone:
    7F6-G5-A2
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Pig,
    Rabbit,
    Human,
    Guinea pig,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Epitope mapped to SQPLPGER in the third intracellular loop (PMID 9822147).
  • Gene ID:
    135
  • Antigen synonyms:
    adenosine A2 receptor|adenosine A2a receptor|adenosine receptor subtype A2a|ADORA2|hA2aR|adenosine receptor A2a|ADORA 2|RDC8
  • Storage buffer:
    PBS
  • Storage temperature:
    Store at 4 °C short term. Aliquot and store at ‒20 °C long term. Avoid freeze-thaw cycles.
  • Concentration:
    1,0 mg/ml
  • Immunogen:
    Full length recombinant human Adenosine A2a R. [UniProt# P29274]
  • Purification:
    Protein G purified
  • Pk:
    100 µl

Specifications

About this item

The Adenosine A2a R Antibody (7F6-G5-A2) from Novus Biologicals is a mouse monoclonal antibody to Adenosine A2a R. This antibody reacts with human, mouse, rat, porcine, canine, guinea pig, rabbit. The Adenosine A2a R Antibody (7F6-G5-A2) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready.

CyTOF to ready

Type: Primary
Antigen: Adenosine A2a R
Clonality: Monoclonal
Clone: 7F6-G5-A2
Conjugation: Unconjugated
Epitope: Epitope mapped to SQPLPGER in the third intracellular loop (PMID 9822147).
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit