3'3'-cGAMP (sodium salt) ≥98%
Supplier: Cayman Chemical
Certificates
About this item
:
Formula:C₂₀H₂₂N₁₀O₁₃P₂ ·2Na
:718.4 g/mol
Storage Temperature:Freezer
Specification Test Results
- Formulation:A crystalline solid
- Purity:≥98%
- Stability:≥ 2 years
Specification Test Results
Frequently Bought Together