2,6-Dichloro-4-fluorobenzoic acid ≥97%
:232275-55-7
:
:C₇H₃Cl₂FO₂
:MFCD23135660
:209.00192 g/mol
Frequently Bought Together
Ethanol, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
: VWR Chemicals
n-Heptane ≥99% (GC (n-heptane basis)), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Ethyl Acetate (BP, Ph. Eur.) pharma grade 1 * 200 L
: PanReac AppliChem
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
: Thermo Fisher Scientific
Tetrahydrofuran 99.8% unstabilised for HPLC
: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
: Thermo Fisher Scientific
Isooctane for UV, IR, HPLC, ACS 1 * 1 L
: PanReac AppliChem