Methyl 4-chloro-2-methyl-pyrazole-3-carboxylate ≥95%
:400877-54-5
:
:C₆H₇ClN₂O₂
:MFCD02253778
:174.58498 g/mol
Frequently Bought Together
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
: VWR Chemicals
Water, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
n-Hexane ≥99.0%, PESTINORM® for capillary GC analysis
: VWR Chemicals
(±)-2-Butanol ≥99% analytical reagent
: VWR Chemicals