High Sensitivity DNA ScreenTape Analysis (ScreenTape and Reagents)
About this item
DNA fragment or library analysis from samples of limited abundance, for samples up to 5000 bp.
- The broad size range allows analysis of DNA fragments or libraries from 35 to 5000 bp
- Results are obtained quickly, in approximately 1 minute per sample
- Detection of DNA samples with limited abundance is possible, for samples down to 5 pg/µl
- Ready-to use ScreenTape devices, pre-packaged reagent and automated data analysis offer an easy to use system with reproducible DNA sizing and quantification results
- Reliable sample quality control is provided for next-generation sequencing (NGS) workflows
- No cross contamination or sample carryover is caused due to the use of single lanes and individual loading tips
- Partially used ScreenTape devices can be used to completion at a later point in time
Agilent's TapeStation High Sensitivity DNA system provides a fast, automated, and flexible solution for samples of limited abundance in many applications. High Sensitivity DNA electrophoresis can be used for quality control of next-generation sequencing (NGS) libraries.
The High Sensitivity DNA ScreenTape assays are suitable for analysis of precious samples. They can separate DNA fragments and libraries up to 5000 base pairs in 1 to 2 minutes per sample. Data output yields objective quantitative and purity measurements from only 2 µl of sample.
Assays are compatible with all Agilent TapeStation instruments.
Note: Some products within these assays contain SYBR™ Green I, which is licensed from Life Technologies Corporation for use in research and development only. SYBR™ is a registered trademark of Life Technologies Corporation.
Caution: For research use only. Not for use in diagnostic procedures.
7 Options Available Below
Product Details & Documents
DNA fragment or library analysis from samples of limited abundance, for samples up to 5000 bp.
- The broad size range allows analysis of DNA fragments or libraries from 35 to 5000 bp
- Results are obtained quickly, in approximately 1 minute per sample
- Detection of DNA samples with limited abundance is possible, for samples down to 5 pg/µl
- Ready-to use ScreenTape devices, pre-packaged reagent and automated data analysis offer an easy to use system with reproducible DNA sizing and quantification results
- Reliable sample quality control is provided for next-generation sequencing (NGS) workflows
- No cross contamination or sample carryover is caused due to the use of single lanes and individual loading tips
- Partially used ScreenTape devices can be used to completion at a later point in time
Agilent's TapeStation High Sensitivity DNA system provides a fast, automated, and flexible solution for samples of limited abundance in many applications. High Sensitivity DNA electrophoresis can be used for quality control of next-generation sequencing (NGS) libraries.
The High Sensitivity DNA ScreenTape assays are suitable for analysis of precious samples. They can separate DNA fragments and libraries up to 5000 base pairs in 1 to 2 minutes per sample. Data output yields objective quantitative and purity measurements from only 2 µl of sample.
Assays are compatible with all Agilent TapeStation instruments.
Note: Some products within these assays contain SYBR™ Green I, which is licensed from Life Technologies Corporation for use in research and development only. SYBR™ is a registered trademark of Life Technologies Corporation.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Product Information
- DetailsAvailabilityPrice
- AGLS5067-5584High sensitivity D1000 ScreenTapeHigh sensitivity D1000 ScreenTapeFor the high sensitivity analysis of DNA from 35 to 1000 bp, includes 7 ScreenTape devices, sufficient for 112 samples, to order with AGLS5067-5585.Log in for availability1 * 1 pezzi372,71 €
- AGLS5067-5585High sensitivity D1000 reagentsHigh sensitivity D1000 reagentsFor the high sensitivity analysis of DNA from 35 to 1000 bp, includes ladder and sample buffer sufficient for 112 samples, to order with AGLS5067-5584Log in for availability1 * 1 pezzi126,00 €
- AGLS5067-5587High sensitivity D1000 ladderHigh sensitivity D1000 ladderFor the high sensitivity analysis of DNA from 35 to 1000 bp, ladder onlyLog in for availability1 * 20 µG66,71 €
- AGLS5067-5603High sensitivity D1000 sample bufferHigh sensitivity D1000 sample bufferFor the high sensitivity analysis of DNA from 35 to 1000 bp, sample buffer onlyLog in for availability1 * 300 µG60,35 €
- AGLS5067-5592High sensitivity D5000 ScreenTapeHigh sensitivity D5000 ScreenTapeFor the high sensitivity analysis of DNA from 100 to 5000 bp, includes 7 ScreenTape devices, sufficient for 105 samples, order with AGLS5067-5593Log in for availability1 * 1 pezzi371,65 €
- AGLS5067-5593High sensitivity D5000 reagentsHigh sensitivity D5000 reagentsFor the high sensitivity analysis of DNA from 100 to 5000 bp, includes ladder and sample buffer sufficient for 105 samples, order with AGLS5067-5592Log in for availability1 * 1 pezzi126,00 €
- AGLS5067-5594High sensitivity D5000 ladderHigh sensitivity D5000 ladderFor the high sensitivity analysis of DNA from 100 to 5000 bp, ladder onlyLog in for availability1 * 20 µG57,18 €