Anti-Nivolumab Mouse Monoclonal Antibody [clone: 6G5]
GENSA01965-40
- Antigen name:Nivolumab
- Clonality:Monoclonal
- Clone:6G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Form:Lyophilized
- Antigen synonyms:Opdivo
- Storage temperature:The lyophilized product remains stable up to 1 year at -20 °C from date of receipt. Upon reconstitution, it can be stored for 2-3 weeks at 2-8 °C or for up to 12 months at -20 °C or below. Avoid freeze/thaw cycles.
- Purification:Protein A affinity column
- Pk:40 µG