
2-Amino-4-cyclopropylbenzoic acid ≥95%
:2640734-77-4
:
:MFCD32862613
:177.20 g/mol
:GHS07
Frequently Bought Together
3-(Chloromethyl)benzoyl chloride 98%
: Thermo Fisher Scientific
2-Butoxyethanol 99%, extra pure
: Thermo Fisher Scientific
Ethyl acetate ≥99.9%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
: Honeywell Chemicals
Ethanol absolute Ph. Eur., J.T.Baker®
: Avantor
2-Methylbutane ≥99%, Reagent Grade
: Honeywell Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Methanol, extra dry 99.9%, AcroSeal®
: Thermo Fisher Scientific
Ethyl acetate ≥99.5% for HPLC
: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
N,N-Dimethylacetamide, anhydrous (max. 0.005% H₂O)
: VWR Chemicals
Chloroform ≥99.5% stabilised
: Honeywell Chemicals