Order Entry
Italy
ContactUsLinkComponent
Industrial nitrile gloves, AlphaTec® 37-300 series
Industrial nitrile gloves, AlphaTec® 37-300 series
Supplier:  Ansell
Certificates
undefined
Industrial nitrile gloves, AlphaTec® 37-300 series
Supplier:  Ansell
Specifications
  • Catalog No:
  • ANSE37-300/7
  • ANSE37-300/8
  • ANSE37-300/11
  • ANSE37-300/10
  • ANSE37-300/9
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Material:
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Cuff style:
  • Gauntlet
  • Gauntlet
  • Gauntlet
  • Gauntlet
  • Gauntlet
  • Sterility:
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Anti-static:
  • +
  • +
  • +
  • +
  • +