Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:1173097-77-2
- Description:Robenidine-d8 hydrochloride
- MDL No.:MFCD10567356
- Synonym(s):1,3-Bis[(4-chlorobenzylidene)amino]guanidine-d8 monohydrochloride
- Melting point:172 °C
- Formula:C₁₅D₈H₅Cl₂N₅·HCl
- Molecular weight:378.71 g/mol
- Description:Robenidine-d8 hydrochloride
- Physical form:Solid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery