Order Entry
Italy
ContactUsLinkComponent
Isobutyl acetate Analytical standard, reference material, Fluka™
Isobutyl acetate Analytical standard, reference material, Fluka™
  HONCR1933-1G
 :  
Isobutyl acetate Analytical standard, reference material, Fluka™
  HONCR1933-1G
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    1 g
  • Standards:
    Analytical standard
  • CAS No.:
    110-19-0
  • Description:
    Isobutyl acetate
  • Storage conditions:
    RT
  • MDL No.:
    MFCD00008932
  • UN No.:
    1213
  • Synonym(s):
    Acetic acid isobutyl ester
  • Formula:
    C₆H₁₂O₂
  • Molecular weight:
    116.16 g/mol
  • Description:
    Isobutyl acetate
  • Flash point:
    22 °C
  • Physical form:
    Liquid

 

 

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery