Order Entry
Italy
Orders LinkContactUsLinkComponent
1-Propanol Analytical standard, reference material, Fluka™

1-Propanol Analytical standard, reference material, Fluka™

1-Propanol, Analytical standard, Glass Bottle, 1 g
Catalog # HONCR1925-1G
Supplier:   Honeywell Chemicals
1-Propanol Analytical standard, reference material, Fluka™
Catalog # HONCR1925-1G
CAS Number:  
1-Propanol Analytical standard, reference material, Fluka™

1-Propanol, Analytical standard, Glass Bottle, 1 g

CAS Number:  
Price
59,49 €
Log in for availability
1 * 1 g
Product Details & Documents

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
Specifications

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    1 g
  • Standards:
    Analytical standard
  • Description:
    1-Propanol
  • UN No.:
    1274
  • Cat. No.:
    HONCR1925-1G
  • Formula:
    C₃H₈O
  • Description:
    1-Propanol
  • Storage conditions:
    RT
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00002941
  • Flash point:
    21 °C
  • Melting point:
    -127 °C
  • Synonym(s):
    Propyl alcohol
  • Molecular weight:
    60.1 g/mol
  • CAS No.:
    71-23-8

Specifications