2-Phenylethanol Analytical standard, reference material, Fluka™
2-Phenylethanol, Analytical standard, Glass Bottle, 1 g
Catalog # HONCR1919-1G
Supplier: Honeywell Chemicals
2-Phenylethanol Analytical standard, reference material, Fluka™
Catalog # HONCR1919-1G
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
- Pack type:Glass Bottle
- Pk:1 g
- Standards:Analytical standard
- Description:2-Phenylethanol
- Cat. No.:HONCR1919-1G
- Formula:C₈H₁₀O
- Description:2-Phenylethanol
- Storage conditions:RT
- Physical form:Liquid
- MDL No.:MFCD00002886
- Flash point:96 °C
- Melting point:-27 °C.(1,013 hPa)
- Synonym(s):2-Phenylethyl alcohol
- Molecular weight:122.16 g/mol
- CAS No.:60-12-8



