Order Entry
Italy
Orders LinkContactUsLinkComponent
Linalool Analytical standard, reference material, Fluka™

Linalool Analytical standard, reference material, Fluka™

Linalool, Analytical standard, Glass Bottle, 1 g
Catalog # HONCR1912-1G
Supplier:   Honeywell Chemicals
Linalool Analytical standard, reference material, Fluka™
Catalog # HONCR1912-1G
CAS Number:  
Linalool Analytical standard, reference material, Fluka™

Linalool, Analytical standard, Glass Bottle, 1 g

CAS Number:  
Price
83,20 €
Log in for availability
1 * 1 g
Product Details & Documents

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
Specifications

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    1 g
  • Standards:
    Analytical standard
  • Description:
    Linalool
  • Cat. No.:
    HONCR1912-1G
  • Formula:
    C₁₀H₁₈O
  • Description:
    Linalool
  • Storage conditions:
    RT
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00008906
  • Flash point:
    77.2 °C.closed cup
  • Melting point:
    < -74 °C
  • Synonym(s):
    (±)-3,7-Dimethyl-1,6-octadien-3-ol
  • Molecular weight:
    154.25 g/mol
  • CAS No.:
    78-70-6

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.