Order Entry
Italy
ContactUsLinkComponent
30 Component Mix for F&B, in ethanol absol. Analytical standard, reference material, Fluka™
30 Component Mix for F&B, in ethanol absol. Analytical standard, reference material, Fluka™
Catalog # HONCR1902-2ML
CAS Number:  
30 Component Mix for F&B, in ethanol absol. Analytical standard, reference material, Fluka™
Catalog # HONCR1902-2ML
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    2 mL
  • Standards:
    Analytical standard
  • CAS No.:
    67-56-1
  • Description:
    30 Component Mix for F&B, in ethanol absol.
  • Storage conditions:
    2-8°C
  • UN No.:
    1170
  • Synonym(s):
    Multi component solution
  • Melting point:
    -115 °C Data refer to the solvent.
  • Description:
    30 Component Mix for F&B, in ethanol absol.
  • Flash point:
    13 °C Ethanol
  • Physical form:
    Liquid

Specifications

About this item

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery