Benalaxyl Analytical standard, reference material, Fluka™
HONCR1858-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:71626-11-4
- Description:Benalaxyl
- Storage conditions:RT
- MDL No.:MFCD00143652
- Synonym(s):Methyl N-(2,6-dimethylphenyl)-N-(phenylacetyl)-DL-alaninate
- Melting point:78 - 80 °C
- Formula:C₂₀H₂₃NO₃
- Molecular weight:325.40 g/mol
- Description:Benalaxyl
- Physical form:Solid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery