Monensin sodium salt hydrate Analytical standard, reference material, Fluka™
Codice catalogo HONCR1805-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:22373-78-0
- Description:Monensin sodium salt hydrate
- Storage conditions:2-8°C
- MDL No.:MFCD00077826
- UN No.:3462
- Molecular weight:692.85 g/mol
- Description:Monensin sodium salt hydrate
- Physical form:Solid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery