Order Entry
Italy
ContactUsLinkComponent
Hydroxylamine solution 50% (w/w) in water for TMT experiments
Corrosive
Health
Irritant
Environment
Hydroxylamine solution 50% (w/w) in water for TMT experiments
Catalog # PIER90115
CAS Number:  
undefined
Corrosive
Health
Irritant
Environment
Hydroxylamine solution 50% (w/w) in water for TMT experiments
Catalog # PIER90115
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • ASSAY/TITRATION:
    45.0 - 55.0% range
  • VISUAL:
    Colorless/clear to slightly yellow liquid

Specification Test Results

About this item

 :
 :33.03 g/mol
Storage Temperature:Ambient
EINECS:232-259-2
UN:3266
ADR:8,III
 :GHS05
 :GHS08
 :GHS07
 :GHS09
CAS Number:7803-49-8
Formula:H₃NO
MDL Number:MFCD00044522