Order Entry
Italy
ContactUsLinkComponent
Estradiol Cypionate System Suitability Mixture, Reference Standard
Estradiol Cypionate System Suitability Mixture, Reference Standard
Catalog # 1252193.
Supplier:  USP
CAS Number:  
Estradiol Cypionate System Suitability Mixture, Reference Standard
Catalog # 1252193.
Supplier:  USP
CAS Number:  
Specifications
  • Pack type:
    Vial
  • Pk:
    10 mg
  • Standards:
    USP reference standard
  • Description:
    Estradiol Cypionate System Suitability Mixture
  • Seq:
    03
Specifications
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
This mixture contains estradiol cypionate, estradiol-9-ene cypionate, estradiol dicypionate, and 4-methylestradiol cypionate
Certifications: US Pharmacopea Reference Standard