Order Entry
Italy
Orders LinkContactUsLinkComponent
Phenytoin, Reference Standards

Phenytoin, Reference Standards

Supplier:   USP
Phenytoin, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1535019.
  • 1535020.
  • 1535008.
  • Description:
  • Phenytoin Related Compound A
  • Phenytoin Related Compound B
  • Phenytoin
  • Synonym(s):
  • 2,2-Diphenylglycine
  • 2,2-Diphenyl-2-ureidoacetic acid
  • CAS No.:
  • 3060-50-2
  • 6802-95-5
  • 57-41-0
  • Formula:
  • C₁₄H₁₃NO₂
  • C₁₅H₁₄N₂O₃

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1535019.
    Phenytoin Related Compound A
    Phenytoin Related Compound A
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 310,00 €
    1 310,00 €
    1 * 50 mg
       1 310,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1535019.
    Description
    Phenytoin Related Compound A
    Synonym(s)
    2,2-Diphenylglycine
    CAS No.
    3060-50-2
    Formula
    C₁₄H₁₃NO₂
  • 1535020.
    Phenytoin Related Compound B
    Phenytoin Related Compound B
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 270,00 €
    1 270,00 €
    1 * 50 mg
       1 270,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1535020.
    Description
    Phenytoin Related Compound B
    Synonym(s)
    2,2-Diphenyl-2-ureidoacetic acid
    CAS No.
    6802-95-5
    Formula
    C₁₅H₁₄N₂O₃
  • 1535008.
    Phenytoin
    Phenytoin
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 403,00 €
    403,00 €
    1 * 200 mg
       403,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1535008.
    Description
    Phenytoin
    Synonym(s)
    CAS No.
    57-41-0
    Formula
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.