Order Entry
Italy
Orders LinkContactUsLinkComponent
Lansoprazole, Reference Standards

Lansoprazole, Reference Standards

Supplier:   USP
Lansoprazole, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1356927.
  • 1356931.
  • 1356916.
  • Description:
  • Lansoprazole Related Compound A
  • Lansoprazole Related Compound B
  • Lansoprazole
  • Synonym(s):
  • 2-({[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridyl]methyl}sulfonyl)benzimidazole
  • 2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)-pyridin-2-yl]methyl]sulfanyl]-1H-benzimidazole monohydrate
  • CAS No.:
  • 131926-99-3
  • 1288338-69-1
  • 103577-45-3
  • Formula:
  • C₁₆H₁₄F₃N₃O₃S

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1356927.
    Lansoprazole Related Compound A
    Lansoprazole Related Compound A
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 300,00 €
    1 300,00 €
    1 * 25 mg
       1 300,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1356927.
    Description
    Lansoprazole Related Compound A
    Synonym(s)
    2-({[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridyl]methyl}sulfonyl)benzimidazole
    CAS No.
    131926-99-3
    Formula
    C₁₆H₁₄F₃N₃O₃S
  • 1356931.
    Lansoprazole Related Compound B
    Lansoprazole Related Compound B
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 300,00 €
    1 300,00 €
    1 * 25 mg
       1 300,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1356931.
    Description
    Lansoprazole Related Compound B
    Synonym(s)
    2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)-pyridin-2-yl]methyl]sulfanyl]-1H-benzimidazole monohydrate
    CAS No.
    1288338-69-1
    Formula
  • 1356916.
    Lansoprazole
    Lansoprazole
    USP reference standard
    Vial
    150 mg
    Log in for availability
    Log in for availability
    1 * 150 mg
    1 * 150 mg - 373,00 €
    373,00 €
    1 * 150 mg
       373,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1356916.
    Description
    Lansoprazole
    Synonym(s)
    CAS No.
    103577-45-3
    Formula
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.